Thermo Scientific Chemicals

4-Fluoro-2,6-dimethylaniline, 97%, Thermo Scientific Chemicals

Catalog number: B24965.03
1 g, Each
Thermo Scientific Chemicals

4-Fluoro-2,6-dimethylaniline, 97%, Thermo Scientific Chemicals

Catalog number: B24965.03
1 g, Each
Quantity
Catalog number: B24965.03
also known as B24965-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
392-70-1
IUPAC Name
4-fluoro-2,6-dimethylaniline
Molecular Formula
C8H10FN
InChI Key
MPNDLCBMLBACDD-UHFFFAOYSA-N
SMILES
CC1=CC(F)=CC(C)=C1N
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to yellow or pale orange to orange to dark brown
Form
Liquid
Refractive Index
1.5325-1.5365 @ 20°C

Description

4-Fluoro-2,6-dimethylaniline is used as an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-2,6-dimethylaniline is used as an intermediate.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text