5-Iodo-2,4-dimethoxypyrimidine, 98%, Thermo Scientific Chemicals
5-Iodo-2,4-dimethoxypyrimidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Iodo-2,4-dimethoxypyrimidine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
0.25 g
5 g
Catalog number B24967.03
also known as B24967-03
Price (USD)/ Each
159.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
159.00
Add to cart
5-Iodo-2,4-dimethoxypyrimidine, 98%, Thermo Scientific Chemicals
Catalog numberB24967.03
Price (USD)/ Each
159.00
-
Add to cart
Chemical Identifiers
CAS52522-99-3
IUPAC Name5-iodo-2,4-dimethoxypyrimidine
Molecular FormulaC6H7IN2O2
InChI KeyKNTMOGOYRYYUIE-UHFFFAOYSA-N
SMILESCOC1=NC=C(I)C(OC)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
FormPowder
Assay (GC)≥97.5%
Melting Point (clear melt)68.0-74.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only