9,10-Anthraquinone-2-sulfonic acid sodium salt hydrate forms charge-transfer complexes with 9,10-dimethoxyanthracene-2-sulfonate. It is used as a catalyst in production of alkaline pulping in the soda process. It is used as an in situ photochemical fluorescence probe.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
9,10-Anthraquinone-2-sulfonic acid sodium salt hydrate forms charge-transfer complexes with 9,10-dimethoxyanthracene-2-sulfonate. It is used as a catalyst in production of alkaline pulping in the soda process. It is used as an in situ photochemical fluorescence probe.
Solubility
Soluble in hot water, cold water and insoluble in alcohol or ether.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
Wen-You Li; Xiang-Qun Guo; Jin-Gou Xu; Qing-Zhi Zhu; Yi-Bing Zhao. Determination of DNA using sodium 9,10-anthraquinone-2-sulfonate as an in situ photochemical fluorescence probe. Analytica Chimica Acta.1997, 340, 291-296.
Haruo Inoue; Mitsuhiko Hida. Photochemical Electron Transport from Water Layer to Organic Layer via Sodium Anthraquinone-2-sulfonate by Stepwise Two Photon Excitation. Bulletin of the Chemical Society of Japan.1982, 52, (6), 1880-1885.