p-Thiocresol, 98%, Thermo Scientific Chemicals
p-Thiocresol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

p-Thiocresol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number B24970.36
also known as B24970-36
Price (USD)/ Each
237.65
Online exclusive
264.00 
Save 26.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
237.65
Online exclusive
264.00 
Save 26.35 (10%)
Add to cart
p-Thiocresol, 98%, Thermo Scientific Chemicals
Catalog numberB24970.36
Price (USD)/ Each
237.65
Online exclusive
264.00 
Save 26.35 (10%)
-
Add to cart
Chemical Identifiers
CAS106-45-6
IUPAC Name4-methylbenzene-1-thiol
Molecular FormulaC7H8S
InChI KeyWLHCBQAPPJAULW-UHFFFAOYSA-N
SMILESCC1=CC=C(S)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)≥97.5%
Melting Point (clear melt)39.0-45.0?C
p-Thiocresol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Thiocresol is used as a pharmaceutical intermediate.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents, air.
RUO – Research Use Only

General References:

  1. Stanley J. Cristol; Gordon D. Brindell; James A. Bridged Polycyclic Compounds. V. The Addition of p-Thiocresol to Norbornadiene. The Question of Non-classical Free Radicals. J. Am. Chem. Soc. 1958, 80 (3), 635-640.
  2. Jianing Chen; Weisheng Yang; Kimberly Dick; Knut Deppert; H. Q. Xu; Lars Samuelson and Hongxing Xu. STip-enhanced Raman scattering of p-thiocresol molecules on individual gold nanoparticles. Appl. Phys. Lett. 2008, 92 (9), 093110.