Thermo Scientific Chemicals

Sodium phenoxide, 98%, Thermo Scientific Chemicals

Catalog number: B25002.18
50 g, Each
Thermo Scientific Chemicals

Sodium phenoxide, 98%, Thermo Scientific Chemicals

Catalog number: B25002.18
50 g, Each
Quantity
Catalog number: B25002.18
also known as B25002-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
139-02-6
IUPAC Name
sodium benzenolate
Molecular Formula
C6H5NaO
InChI Key
NESLWCLHZZISNB-UHFFFAOYSA-M
SMILES
[Na+].[O-]C1=CC=CC=C1
Appearance (Color)
White to pale brown
Form
powder
Assay (Non-aqueous acid-base Titration)
> 97.5%

Description

Sodium phenoxide is used as a precursor to aryl ethers and involved in the preparation of phenyl ethers and metal phenolates. It serves as an antiseptic and used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium phenoxide is used as a precursor to aryl ethers and involved in the preparation of phenyl ethers and metal phenolates. It serves as an antiseptic and used in organic synthesis.

Solubility
Very soluble in water, soluble in alcoholSoluble in water, acetone and alcohol.

Notes
Hygroscopic and air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text