Thermo Scientific Chemicals

Creatine monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: B25009.36
500 g, Each
Thermo Scientific Chemicals

Creatine monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: B25009.36
500 g, Each
Quantity
Catalog number: B25009.36
also known as B25009-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6020-87-7
IUPAC Name
2-(N-methylcarbamimidamido)acetic acid hydrate
Molecular Formula
C4H11N3O3
InChI Key
MEJYXFHCRXAUIL-UHFFFAOYSA-N
SMILES
O.CN(CC(O)=O)C(N)=N
Form
Powder or crystalline powder
Loss on Drying
6.43-17.09% (105°C, 0.5-1.5 waters)
Appearance (Color)
White to cream to yellow
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

Creatine monohydrate is involved in rapid ATP production primarily in skeletal muscle tissue via the action of creatine kinase(s). It may be used as a supplement to study its uptake mechanism and metabolism of action. It is used in the treatment of neuromuscular diseases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (17 g/L) at 20°C. Insoluble in ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text