Thermo Scientific Chemicals

3-Chloropropyl acetate, 97+%, Thermo Scientific Chemicals

Catalog number: B25013.22
100 g, Each
Thermo Scientific Chemicals

3-Chloropropyl acetate, 97+%, Thermo Scientific Chemicals

Catalog number: B25013.22
100 g, Each
Quantity
Catalog number: B25013.22
also known as B25013-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
628-09-1
IUPAC Name
3-chloropropyl acetate
Molecular Formula
C5H9ClO2
InChI Key
KPOHQIPNNIMWRL-UHFFFAOYSA-N
SMILES
CC(=O)OCCCCl
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.4280-1.4320 @ 20?C

Description

3-Chloropropyl acetate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agro chemicals, dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloropropyl acetate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agro chemicals, dyestuff.

Solubility
Soluble in water (partly).

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text