3-Chloropropyl acetate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agro chemicals, dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Chloropropyl acetate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agro chemicals, dyestuff.
Solubility
Soluble in water (partly).
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only
Donald C. Dittmer; Walter R. Hertler; Herbert Winicov. Mechanism of Trimethylene Oxide Formation from 3-Chloropropyl Acetate. J. Am. Chem. Soc. 1957, 79 (16), 4431-4435.
Franz Sondheimer; R. B. Woodward. A Synthesis of 1,3-Butylene Oxide. J. Am. Chem. Soc. 1953, 75 (21), 5438-5439.