Thermo Scientific Chemicals

Chlorocyclopentane, 99%, Thermo Scientific Chemicals

Catalog number: B25023.22
100 g, Each
Thermo Scientific Chemicals

Chlorocyclopentane, 99%, Thermo Scientific Chemicals

Catalog number: B25023.22
100 g, Each
Quantity
Catalog number: B25023.22
also known as B25023-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
930-28-9
IUPAC Name
chlorocyclopentane
Molecular Formula
C5H9Cl
InChI Key
NDTCXABJQNJPCF-UHFFFAOYSA-N
SMILES
ClC1CCCC1
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.4500-1.4530 @ 20?C
Form
Liquid

Description

Chlorocyclopentane was used in the preparation of diastereomeric activation products. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorocyclopentane was used in the preparation of diastereomeric activation products. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water. 760 mg/L.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text