2,5-Dihydro-2,5-dimethoxyfuran, cis + trans, 99%, Thermo Scientific Chemicals
2,5-Dihydro-2,5-dimethoxyfuran, cis + trans, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,5-Dihydro-2,5-dimethoxyfuran, cis + trans, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
100 g
250 g
500 g
Catalog number B25028.0B
also known as B25028-0B
Price (USD)/ Each
508.00
-
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)/ Each
508.00
Add to cart
2,5-Dihydro-2,5-dimethoxyfuran, cis + trans, 99%, Thermo Scientific Chemicals
Catalog numberB25028.0B
Price (USD)/ Each
508.00
-
Add to cart
Chemical Identifiers
CAS332-77-4
IUPAC Name2,5-dimethoxy-2,5-dihydrofuran
Molecular FormulaC6H10O3
InChI KeyWXFWXFIWDGJRSC-UHFFFAOYNA-N
SMILESCOC1OC(OC)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.4335-1.4365 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only