Thermo Scientific Chemicals

Imidazole-4-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: B25050.03
1 g, Each
Thermo Scientific Chemicals

Imidazole-4-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: B25050.03
1 g, Each
Quantity
Catalog number: B25050.03
also known as B25050-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3034-50-2
IUPAC Name
1H-imidazole-5-carbaldehyde
Molecular Formula
C4H4N2O
InChI Key
ZQEXIXXJFSQPNA-UHFFFAOYSA-N
SMILES
O=CC1=CN=CN1
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥96.0%
Melting Point (clear melt)
168.0-178.0?C
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0%
Appearance (Color)
Cream to yellow

Description

Imidazole-4-carboxaldehyde is a 4-formyl derivative of imidazole used in the preparation of C17,20-lyase inhibitor for the treatment of androgen-dependent prostate cancer. It is also used in the synthesis of other biologically active compounds such as antimalarial drugs, fabrication of colorimetric chemosensor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Imidazole-4-carboxaldehyde is a 4-formyl derivative of imidazole used in the preparation of C17,20-lyase inhibitor for the treatment of androgen-dependent prostate cancer. It is also used in the synthesis of other biologically active compounds such as antimalarial drugs, fabrication of colorimetric chemosensor.

Solubility
Soluble in DMSO, methanol.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text