Imidazole-4-carboxaldehyde is a 4-formyl derivative of imidazole used in the preparation of C17,20-lyase inhibitor for the treatment of androgen-dependent prostate cancer. It is also used in the synthesis of other biologically active compounds such as antimalarial drugs, fabrication of colorimetric chemosensor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Imidazole-4-carboxaldehyde is a 4-formyl derivative of imidazole used in the preparation of C17,20-lyase inhibitor for the treatment of androgen-dependent prostate cancer. It is also used in the synthesis of other biologically active compounds such as antimalarial drugs, fabrication of colorimetric chemosensor.
Solubility
Soluble in DMSO, methanol.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
RUO – Research Use Only
General References:
- Seon-Yeong Gwon; Sung-Hoon Kim. Anion sensing and F--induced reversible photoreaction of D-π-A type dye containing imidazole moiety as donor. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2014, 117, 810-813.
- K Honda; Y Nishijima. Convenient synthesis of alkyl esters of urocanic acid. Journal of Pharmaceutical Sciences. 1981, 70 (1), 98-99.