Thermo Scientific Chemicals

4-Azabenzimidazole, 98%, Thermo Scientific Chemicals

Catalog number: B25051.14
25 g, Each
Thermo Scientific Chemicals

4-Azabenzimidazole, 98%, Thermo Scientific Chemicals

Catalog number: B25051.14
25 g, Each
Quantity
Catalog number: B25051.14
also known as B25051-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
273-21-2
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Melting Point (clear melt)
142.5-148.5°C
Appearance (Color)
Pale brown to brown or pale grey to grey
Assay (GC)
≥97.5%

Description

4-Azabenzimidazole is used in the preparation of imidazo[4,5-b]pyridine 4-oxide. It is also used as a reagent to prepare benzoylmethyl-substituted azoles which are inhibitors of nitric oxide synthase with antioxidant properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Azabenzimidazole is used in the preparation of imidazo[4,5-b]pyridine 4-oxide. It is also used as a reagent to prepare benzoylmethyl-substituted azoles which are inhibitors of nitric oxide synthase with antioxidant properties.

Solubility
Soluble in dimethylformamide.

Notes
Incompatible with strong oxidizing agent and strong acid.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text