Thermo Scientific Chemicals

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, 97%, Thermo Scientific Chemicals

Catalog number: B25057.06
5 g, Each
Thermo Scientific Chemicals

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, 97%, Thermo Scientific Chemicals

Catalog number: B25057.06
5 g, Each
Quantity
Catalog number: B25057.06
also known as B25057-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1892-57-5
IUPAC Name
({[3-(dimethylamino)propyl]imino}methylidene)(ethyl)amine
Molecular Formula
C8H17N3
InChI Key
LMDZBCPBFSXMTL-UHFFFAOYSA-N
SMILES
CCN=C=NCCCN(C)C
Refractive Index
1.4590-1.4630 @ 20°C
Assay (GC)
≥96.0%
Form
Liquid
Appearance (Color)
Clear colorless to yellow
Identification (FTIR)
Conforms

Description

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide is used as a carboxyl activating agent and activate phosphate groups in phospho mono and di esters. It is used in peptide synthesis, 3'-amino-3'-deoxyadenosine-5'-di- and triphosphates and in the preparation of antibodies like immunoconjugates. It plays a vital role for immobilization of large biomolecules in association with N-hydroxysuccinimide. It is also used in the acylation of phosphoranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide is used as a carboxyl activating agent and activate phosphate groups in phospho mono and di esters. It is used in peptide synthesis, 3′-amino-3′-deoxyadenosine-5′-di- and triphosphates and in the preparation of antibodies like immunoconjugates. It plays a vital role for immobilization of large biomolecules in association with N-hydroxysuccinimide. It is also used in the acylation of phosphoranes.

Solubility
Soluble in water.

Notes
Air sensitive. Incompatible with strong acids and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text