Thermo Scientific Chemicals

Ethyl cyclohexylideneacetate, 98%, Thermo Scientific Chemicals

Catalog number: B25073.06
5 g, Each
Thermo Scientific Chemicals

Ethyl cyclohexylideneacetate, 98%, Thermo Scientific Chemicals

Catalog number: B25073.06
5 g, Each
Quantity
Catalog number: B25073.06
also known as B25073-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1552-92-7
IUPAC Name
ethyl 2-cyclohexylideneacetate
Molecular Formula
C10H16O2
InChI Key
MCWDXHYYYNGYGK-UHFFFAOYSA-N
SMILES
CCOC(=O)C=C1CCCCC1
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.4770-1.4810 @ 20?C

Description

Ethyl cyclohexylideneacetate is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl cyclohexylideneacetate is used as pharmaceutical intermediates.

Solubility
Soluble in dichloromethane, ethyl acetate.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text