2,6-Difluorotoluene, 98%, Thermo Scientific Chemicals
2,6-Difluorotoluene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Difluorotoluene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number B25076.14
also known as B25076-14
Price (USD)/ Each
253.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
253.00
Add to cart
2,6-Difluorotoluene, 98%, Thermo Scientific Chemicals
Catalog numberB25076.14
Price (USD)/ Each
253.00
-
Add to cart
Chemical Identifiers
CAS443-84-5
IUPAC Name1,3-difluoro-2-methylbenzene
Molecular FormulaC7H6F2
InChI KeyMZLSNIREOQCDED-UHFFFAOYSA-N
SMILESCC1=C(F)C=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.4510-1.4550 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only