4,4'-Dimethylbiphenyl is used in the preparation of biphenyl-4,4’-dicarboxylic acid, which acts as a monomer utilized in the synthesis of linear long chain polymer through polycondensation reaction. It is used to prepare 4-(4'-methylphenyl)benzaldehyde by selective photoxygenation reaction using 9-phenyl-10-methylacridinium perchlorate as a catalyst under visible light irradiation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,4′-Dimethylbiphenyl is used in the preparation of biphenyl-4,4’-dicarboxylic acid, which acts as a monomer utilized in the synthesis of linear long chain polymer through polycondensation reaction. It is used to prepare 4-(4′-methylphenyl)benzaldehyde by selective photoxygenation reaction using 9-phenyl-10-methylacridinium perchlorate as a catalyst under visible light irradiation.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
- Guo, M.; Qi, L.; Lv, M.; Zhou, X.; Liang, H.; Chen, S. Homocoupling of Arylboronic Acids Catalyzed by a Simple Hydrophilic Palladium(II) Complex in Aqueous Media. Lett. Org. Chem. 2015, 12 (3), 205-209.
- Raul, P. K.; Mahanta, A.; Bora, U.; Thakur, A. J.; Veer, V. In water homocoupling of arylboronic acids using nano-rod shaped and reusable copper oxide(II) catalyst at room temperature. Tetrahedron Lett. 2015, 56 (51), 7069-7073.