3-Bromothiophenol, 97%, Thermo Scientific Chemicals
3-Bromothiophenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromothiophenol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number B25092.06
also known as B25092-06
Price (USD)/ Each
111.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
111.00
Add to cart
3-Bromothiophenol, 97%, Thermo Scientific Chemicals
Catalog numberB25092.06
Price (USD)/ Each
111.00
-
Add to cart
Chemical Identifiers
CAS6320-01-0
IUPAC Name3-bromobenzene-1-thiol
Molecular FormulaC6H5BrS
InChI KeyHNGQQUDFJDROPY-UHFFFAOYSA-N
SMILESSC1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only