4-Pyridineacetonitrile hydrochloride, 98%, Thermo Scientific Chemicals
4-Pyridineacetonitrile hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Pyridineacetonitrile hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number B25095.06
also known as B25095-06
Price (USD)/ Each
173.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
173.00
Add to cart
4-Pyridineacetonitrile hydrochloride, 98%, Thermo Scientific Chemicals
Catalog numberB25095.06
Price (USD)/ Each
173.00
-
Add to cart
Chemical Identifiers
CAS92333-25-0
IUPAC Name2-(pyridin-4-yl)acetonitrile
Molecular FormulaC7H6N2
InChI KeyBMVSAKPRNWZCPG-UHFFFAOYSA-N
SMILESN#CCC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark red to red
FormPowder
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only