Thermo Scientific Chemicals

4-Iodobenzonitrile, 98%, Thermo Scientific Chemicals

Catalog number: B25104.06
5 g, Each
Thermo Scientific Chemicals

4-Iodobenzonitrile, 98%, Thermo Scientific Chemicals

Catalog number: B25104.06
5 g, Each
Quantity
Catalog number: B25104.06
also known as B25104-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3058-39-7
Appearance (Color)
White to pale cream to cream to yellow to pale brown
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
123.0-130.0?C

Description

4-Iodobenzonitrile is used in organic synthesis. It is used as catalytic agent, petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzonitrile is used in organic synthesis. It is used as catalytic agent, petrochemical additive.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in the dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text