4-Iodobenzonitrile is used in organic synthesis. It is used as catalytic agent, petrochemical additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Iodobenzonitrile is used in organic synthesis. It is used as catalytic agent, petrochemical additive.
Solubility
Insoluble in water.
Notes
Light Sensitive. Store in the dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only
Gautam R. Desiraju; Richard L. Harlow. Cyano-halogen interactions and their role in the crystal structures of the 4-halobenzonitriles. Journal of the American Chemical Society.1989, 111, (17), 6757-6764.