Methyl 6-bromo-2-naphthoate, 98%, Thermo Scientific Chemicals
Methyl 6-bromo-2-naphthoate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 6-bromo-2-naphthoate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number B25106.14
also known as B25106-14
Price (USD)/ Each
214.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
214.00
Add to cart
Methyl 6-bromo-2-naphthoate, 98%, Thermo Scientific Chemicals
Catalog numberB25106.14
Price (USD)/ Each
214.00
-
Add to cart
Chemical Identifiers
CAS33626-98-1
IUPAC Namemethyl 6-bromonaphthalene-2-carboxylate
Molecular FormulaC12H9BrO2
InChI KeyJEUBRLPXJZOGPX-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C2C=C(Br)C=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥97.5%
Melting Point (clear melt)122.0-129.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only