Thermo Scientific Chemicals

3-Methyl-3-oxetanemethanol, 97 %, Thermo Scientific Chemicals

Référence: B25111.22
100 g, Each
Thermo Scientific Chemicals

3-Methyl-3-oxetanemethanol, 97 %, Thermo Scientific Chemicals

Référence: B25111.22
100 g, Each
Quantité
Référence: B25111.22
également connu sous le numéro B25111-22
Prix (EUR)
Quantité
-

Identifiants chimiques

CAS
3143-02-0
IUPAC Name
(3-methyloxetan-3-yl)methanol
Molecular Formula
C5H10O2
InChI Key
NLQMSBJFLQPLIJ-UHFFFAOYSA-N
SMILES
CC1(CO)COC1
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Refractive Index
1.4435-1.4485 @ 20?C

Description

3-Methyl-3-oxetanemethanol has been used in the preparation of star-shaped copolymer consisting of a hyperbranched poly(3-methyl-3-oxetanemethanol) core and polytetrahydrofuran arms, pyridyl disulfide-functionalized cyclic carbonate monomer, required for the synthesis of of functional poly(ε-caprolactone) containing pendant pyridyl disulphide groups and δ-lactams. It is used to synthesize octahydroindole alkaloids and γ-butenolides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-méthyl-3-oxetaneméthanol : pour préparer des copolymères en forme d’étoile composés d’un noyau poly(3-méthyl-3-oxetaneméthanol) hyperramifié et de bras polytétrahydrofurannes, monomère de carbonate cyclique disulfure fonctionnalisé pyridyle, requis pour la synthèse de poly(ε-caprolactone) fonctionnel contenant des groupes de disulfures de pyridyles pendants et δ-lactames. Pour synthétiser des alcaloïdes octahydroindoles et γ-buténolides.

Solubilité
Soluble dans le chloroforme et le méthanol.

Remarques
Protège de la chaleur. Conserver à l’abri des oxydants. Conserver à 4°C.
RUO – Research Use Only

Figures

Documentation et téléchargements

Certificats

    Foire aux questions (FAQ)

    Citations et références

    Search citations by name, author, journal title or abstract text

    Sécurité et manipulation


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute dermal toxicity
    Category 4
    Acute Inhalation Toxicity - Vapors
    Category 4
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Skin Sensitization
    Category 1
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 + H312 + H332 - Harmful if swallowed, in contact with skin or if inhaled

    H315 - Causes skin irritation

    H317 - May cause an allergic skin reaction

    H319 - Causes serious eye irritation

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P337 + P313 - If eye irritation persists: Get medical advice/attention