Pyridine-2,3-dicarboxylic anhydride, 98%, Thermo Scientific Chemicals
Pyridine-2,3-dicarboxylic anhydride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pyridine-2,3-dicarboxylic anhydride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B25118.06
also known as B25118-06
Price (USD)/ Each
44.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
44.00
Add to cart
Pyridine-2,3-dicarboxylic anhydride, 98%, Thermo Scientific Chemicals
Catalog numberB25118.06
Price (USD)/ Each
44.00
-
Add to cart
Chemical Identifiers
CAS699-98-9
IUPAC Name5H,7H-furo[3,4-b]pyridine-5,7-dione
Molecular FormulaC7H3NO3
InChI KeyMCQOWYALZVKMAR-UHFFFAOYSA-N
SMILESO=C1OC(=O)C2=NC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Appearance (Color)White to pale cream
FormPowder or granules
Pyridine-2,3-dicarboxylic anhydride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine-2,3-dicarboxylic anhydride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Optimal solubility in water.

Notes
Moisture sensitive. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.
RUO – Research Use Only