Thermo Scientific Chemicals

(R)-(-)-1,3-Butanediol, 98%, Thermo Scientific Chemicals

Catalog number: B25124.06
5 g, Each
Thermo Scientific Chemicals

(R)-(-)-1,3-Butanediol, 98%, Thermo Scientific Chemicals

Catalog number: B25124.06
5 g, Each
Quantity
Catalog number: B25124.06
also known as B25124-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6290-03-5
IUPAC Name
butane-1,3-diol
Molecular Formula
C4H10O2
InChI Key
PUPZLCDOIYMWBV-UHFFFAOYNA-N
SMILES
CC(O)CCO
Appearance (Color)
Clear colorless to pale yellow
Form
Viscous liquid
Assay (GC)
≥97.5%
Refractive Index
1.4385-1.4425 @ 20?C

Description

(R)-(-)-1,3-Butanediol is used as a solvent for food flavoring agents and is a co-monomer used in certain polyurethane and polyester resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-1,3-Butanediol is used as a solvent for food flavoring agents and is a co-monomer used in certain polyurethane and polyester resins.

Solubility
Fully miscible in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text