4,4'-Dibromooctafluorobiphenyl, 99%, Thermo Scientific Chemicals
4,4'-Dibromooctafluorobiphenyl, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,4'-Dibromooctafluorobiphenyl, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B25126.03
also known as B25126-03
Price (USD)/ Each
83.10
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
83.10
Add to cart
4,4'-Dibromooctafluorobiphenyl, 99%, Thermo Scientific Chemicals
Catalog numberB25126.03
Price (USD)/ Each
83.10
-
Add to cart
Chemical Identifiers
CAS10386-84-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Melting Point (clear melt)111.0-117.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only