Thermo Scientific Chemicals

N-(6-Bromohexyl)phthalimide, 97%, Thermo Scientific Chemicals

Catalog number: B25128.06
5 g, Each
Thermo Scientific Chemicals

N-(6-Bromohexyl)phthalimide, 97%, Thermo Scientific Chemicals

Catalog number: B25128.06
5 g, Each
Quantity
Catalog number: B25128.06
also known as B25128-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
24566-79-8
IUPAC Name
2-(6-bromohexyl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular Formula
C14H16BrNO2
InChI Key
OAZFTIPKNPTDIO-UHFFFAOYSA-N
SMILES
BrCCCCCCN1C(=O)C2=CC=CC=C2C1=O
Appearance (Color)
White to pale cream to pale gray
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Melting Point (clear melt)
51.0-60.0?C

Description

N-(6-Bromohexyl)phthalimide is used as a pharmaceutical intermediate and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(6-Bromohexyl)phthalimide is used as a pharmaceutical intermediate and in organic synthesis.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text