N-(6-Bromohexyl)phthalimide, 97%, Thermo Scientific Chemicals
N-(6-Bromohexyl)phthalimide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-(6-Bromohexyl)phthalimide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number B25128.22
also known as B25128-22
Price (USD)/ Each
1,712.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
1,712.00
Add to cart
N-(6-Bromohexyl)phthalimide, 97%, Thermo Scientific Chemicals
Catalog numberB25128.22
Price (USD)/ Each
1,712.00
-
Add to cart
Chemical Identifiers
CAS24566-79-8
IUPAC Name2-(6-bromohexyl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC14H16BrNO2
InChI KeyOAZFTIPKNPTDIO-UHFFFAOYSA-N
SMILESBrCCCCCCN1C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale gray
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)51.0-60.0?C
N-(6-Bromohexyl)phthalimide is used as a pharmaceutical intermediate and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(6-Bromohexyl)phthalimide is used as a pharmaceutical intermediate and in organic synthesis.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Lu, X.; Li, X.; Cao, Y.; Schultz, A.; Wang, J. L.; Moorefield, C. N.; Wesdemiotis, C.; Cheng, S. Z.; Newkome, G. R. Self-Assembly of a Supramolecular, Three-Dimensional, Spoked, Bicycle-like Wheel. Ange. Chem. Int. Edi. 2013, 52 (30), 7728-7731.
  2. Halby, L.; Champion, C.; Sénamaud-Beaufort, C.; Ajjan, S.; Drujon, T.; Rajavelu, A.; Ceccaldi, A.; Jurkowska, R.; Lequin, O.; Nelson, W. G. Rapid synthesis of new DNMT inhibitors derivatives of procainamide. Chembiochem 2012, 13 (1), 157-165.