Thermo Scientific Chemicals

Potassium D-gluconate, 99%, Thermo Scientific Chemicals

Catalog number: B25135.30
250 g, Each
Thermo Scientific Chemicals

Potassium D-gluconate, 99%, Thermo Scientific Chemicals

Catalog number: B25135.30
250 g, Each
Quantity
Catalog number: B25135.30
also known as B25135-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
299-27-4
IUPAC Name
potassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
Molecular Formula
C6H11KO7
InChI Key
HLCFGWHYROZGBI-JJKGCWMISA-M
SMILES
[K+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5% (dry wt. basis)
Comment
Reducing substances ≤ 1.0%
Loss on Drying
≤ 3.0% (105?C, 4h)

Description

Potassium D-gluconate widely used as sequestering compounds for metal ions in alkaline systems. It is used in a newly developed process entitled ARS for the prevention of shrinkage in dyed wool fabric. Potassium gluconate has been shown to elevate lactate production, and ATP content, as well as induce the Hemoglobin (methemoglobin) reduction rate in rat erythrocytes. It is used in cell culture and in large scale applications such as the cleaning of metal surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium D-gluconate widely used as sequestering compounds for metal ions in alkaline systems. It is used in a newly developed process entitled ARS for the prevention of shrinkage in dyed wool fabric. Potassium gluconate has been shown to elevate lactate production, and ATP content, as well as induce the Hemoglobin (methemoglobin) reduction rate in rat erythrocytes. It is used in cell culture and in large scale applications such as the cleaning of metal surfaces.

Solubility
Soluble in water. (50 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text