N-Benzylidenemethylamine, 98+%, Thermo Scientific Chemicals
N-Benzylidenemethylamine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Benzylidenemethylamine, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number B25161.22
also known as B25161-22
Price (USD)/ Each
388.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
388.00
Add to cart
N-Benzylidenemethylamine, 98+%, Thermo Scientific Chemicals
Catalog numberB25161.22
Price (USD)/ Each
388.00
-
Add to cart
Chemical Identifiers
CAS622-29-7
IUPAC Namemethyl(phenylmethylidene)amine
Molecular FormulaC8H9N
InChI KeyHXTGGPKOEKKUQO-UHFFFAOYSA-N
SMILESCN=CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless to yellow
Assay (GC)>98.0%
Refractive Index1.5500-1.5540
Formliquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only