Methyl isoxazole-5-carboxylate, 97%, Thermo Scientific Chemicals
Methyl isoxazole-5-carboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl isoxazole-5-carboxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number B25169.06
also known as B25169-06
Price (USD)/ Each
423.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
423.00
Add to cart
Methyl isoxazole-5-carboxylate, 97%, Thermo Scientific Chemicals
Catalog numberB25169.06
Price (USD)/ Each
423.00
-
Add to cart
Chemical Identifiers
CAS15055-81-9
IUPAC Namemethyl 1,2-oxazole-5-carboxylate
Molecular FormulaC5H5NO3
InChI KeyILPCPKZAAQXHKL-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=NO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)44.0-53.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only