5-Iodo-1,3-dimethyluracil, 99%, Thermo Scientific Chemicals
5-Iodo-1,3-dimethyluracil, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Iodo-1,3-dimethyluracil, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B25173.03
also known as B25173-03
Price (USD)/ Each
71.20
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
71.20
Add to cart
5-Iodo-1,3-dimethyluracil, 99%, Thermo Scientific Chemicals
Catalog numberB25173.03
Price (USD)/ Each
71.20
-
Add to cart
Chemical Identifiers
CAS40738-83-8
IUPAC Name5-iodo-1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC6H7IN2O2
InChI KeyXUUVRIXNFFFPCM-UHFFFAOYSA-N
SMILESCN1C=C(I)C(=O)N(C)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Melting Point (clear melt)227.0-233.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only