Thermo Scientific Chemicals

2-(Trifluoromethyl)phenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: B25182.06
5 g, Each
Thermo Scientific Chemicals

2-(Trifluoromethyl)phenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: B25182.06
5 g, Each
Quantity
Catalog number: B25182.06
also known as B25182-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3107-34-4
IUPAC Name
[2-(trifluoromethyl)phenyl]hydrazine
Molecular Formula
C7H7F3N2
InChI Key
JSWQDLBFVSTSIW-UHFFFAOYSA-N
SMILES
NNC1=CC=CC=C1C(F)(F)F
Appearance (Color)
White to cream or pale yellow
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text