2-(Trifluoromethyl)phenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals
2-(Trifluoromethyl)phenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Trifluoromethyl)phenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number B25182.14
also known as B25182-14
Price (USD)/ Each
326.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
326.00
Add to cart
2-(Trifluoromethyl)phenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals
Catalog numberB25182.14
Price (USD)/ Each
326.00
-
Add to cart
Chemical Identifiers
CAS3107-34-4
IUPAC Name[2-(trifluoromethyl)phenyl]hydrazine
Molecular FormulaC7H7F3N2
InChI KeyJSWQDLBFVSTSIW-UHFFFAOYSA-N
SMILESNNC1=CC=CC=C1C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only