Thermo Scientific Chemicals

6(5H)-Phenanthridinone, 96%, Thermo Scientific Chemicals

Catalog number: B25187.03
1 g, Each
Thermo Scientific Chemicals

6(5H)-Phenanthridinone, 96%, Thermo Scientific Chemicals

Catalog number: B25187.03
1 g, Each
Quantity
Catalog number: B25187.03
also known as B25187-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1015-89-0
IUPAC Name
5,6-dihydrophenanthridin-6-one
Molecular Formula
C13H9NO
InChI Key
RZFVLEJOHSLEFR-UHFFFAOYSA-N
SMILES
O=C1NC2=CC=CC=C2C2=CC=CC=C12
Form
Powder
Appearance (Color)
Cream to pale brown
Assay (HPLC)
≥95.0%
Melting Point (clear melt)
288.0-298.0?C

Description

6(5H)-Phenanthridinone is used as a probe into the role of PARP in cellular response to irradiation. Treatment of cells with 6(5H)-Phenanthridinone and various DNA-damaging agents showed a resistance to apoptosis. It acts as a reactant in the synthesis of 5,6-dihydrophenanthridine sulfonamides, oxidative coupling with diphenylacetylene, direct copper acetate-catalyzed N-cyclopropylation of cyclic amides. It is used as HIV-1 integrase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6(5H)-Phenanthridinone is used as a probe into the role of PARP in cellular response to irradiation. Treatment of cells with 6(5H)-Phenanthridinone and various DNA-damaging agents showed a resistance to apoptosis. It acts as a reactant in the synthesis of 5,6-dihydrophenanthridine sulfonamides, oxidative coupling with diphenylacetylene, direct copper acetate-catalyzed N-cyclopropylation of cyclic amides. It is used as HIV-1 integrase inhibitor.

Solubility
Soluble in DMSO (5 mg/ml). Insoluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text