Thermo Scientific Chemicals

4-Phenylazophenol, 98%, Thermo Scientific Chemicals

Catalog number: B25198.22
100 g, Each
Thermo Scientific Chemicals

4-Phenylazophenol, 98%, Thermo Scientific Chemicals

Catalog number: B25198.22
100 g, Each
Quantity
Catalog number: B25198.22
also known as B25198-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1689-82-3
IUPAC Name
4-(2-phenylhydrazin-1-ylidene)cyclohexa-2,5-dien-1-one
Molecular Formula
C12H10N2O
InChI Key
JTSBGMZPPPULTA-UHFFFAOYSA-N
SMILES
O=C1C=CC(C=C1)=NNC1=CC=CC=C1
Appearance (Color)
Yellow to orange to brown or green to brown
Form
Powder or crystalline powder
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
151-157°C

Description

4-Phenylazophenol is used to varnish, grease, paraffin wax, resin, soap kind of coloring. It is used in printing/solvent based writing inks. It is used for providing transparent metallic effects like gold imitation provided on metalized polyester films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenylazophenol is used to varnish, grease, paraffin wax, resin, soap kind of coloring. It is used in printing/solvent based writing inks. It is used for providing transparent metallic effects like gold imitation provided on metalized polyester films.

Solubility
Soluble in acetone, ethanol, benzene and ether. Insoluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text