2,4-Dimethyl-6-nitrophenol, 98%, Thermo Scientific Chemicals
2,4-Dimethyl-6-nitrophenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-Dimethyl-6-nitrophenol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number B25205.22
also known as B25205-22
Price (USD)/ Each
828.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
828.00
Add to cart
2,4-Dimethyl-6-nitrophenol, 98%, Thermo Scientific Chemicals
Catalog numberB25205.22
Price (USD)/ Each
828.00
-
Add to cart
Chemical Identifiers
CAS14452-34-7
IUPAC Name2,4-dimethyl-6-nitrophenol
Molecular FormulaC8H9NO3
InChI KeyKJRCHILWKQLEBC-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(O)C(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)66.0-72.0?C
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only