Thermo Scientific Chemicals

Quinoline N-oxide hydrate, 97% (dry wt.), water ca 20%, Thermo Scientific Chemicals

Catalog number: B25209.06
5 g, Each
Thermo Scientific Chemicals

Quinoline N-oxide hydrate, 97% (dry wt.), water ca 20%, Thermo Scientific Chemicals

Catalog number: B25209.06
5 g, Each
Quantity
Catalog number: B25209.06
also known as B25209-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
64201-64-5
IUPAC Name
quinolin-1-ium-1-olate
Molecular Formula
C9H7NO
InChI Key
GIIWGCBLYNDKBO-UHFFFAOYSA-N
SMILES
[O-][N+]1=CC=CC2=CC=CC=C12
Appearance (Color)
Cream to pale brown
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥96.0% (dry wt. basis)
Water Content (Karl Fischer Titration)
≤23.68%
Melting Point (clear melt)
45.0-55.0?C

Description

Quinoline N-oxide hydrate is used in quantitative determination of nitrones using trifluoroacetic anhydride-sodium iodide reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinoline N-oxide hydrate is used in quantitative determination of nitrones using trifluoroacetic anhydride-sodium iodide reagent.

Notes
Hygroscopic. Store away from air and oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text