Thermo Scientific Chemicals

2-Ethyl-6-methylphenol, 97%, Thermo Scientific Chemicals

Catalog number: B25213.06
5 g, Each
Thermo Scientific Chemicals

2-Ethyl-6-methylphenol, 97%, Thermo Scientific Chemicals

Catalog number: B25213.06
5 g, Each
Quantity
Catalog number: B25213.06
also known as B25213-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1687-64-5
IUPAC Name
2-ethyl-6-methylphenol
Molecular Formula
C9H12O
InChI Key
CIRRFAQIWQFQSS-UHFFFAOYSA-N
SMILES
CCC1=CC=CC(C)=C1O
Appearance (Color)
Clear colorless to yellow to orange
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.5275-1.5325 @ 20?C

Description

2-Ethyl-6-methylphenol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethyl-6-methylphenol is used as pharmaceutical intermediate.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text