Thermo Scientific Chemicals

Itaconic anhydride, 97%, Thermo Scientific Chemicals

Catalog number: B25216.36
500 g, Each
Thermo Scientific Chemicals

Itaconic anhydride, 97%, Thermo Scientific Chemicals

Catalog number: B25216.36
500 g, Each
Quantity
Catalog number: B25216.36
also known as B25216-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2170-03-8
IUPAC Name
3-methylideneoxolane-2,5-dione
Molecular Formula
C5H4O3
InChI Key
OFNISBHGPNMTMS-UHFFFAOYSA-N
SMILES
C=C1CC(=O)OC1=O
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Melting Point (clear melt)
65.0-72.0?C

Description

Itaconic anhydride is used in immobilization of β-glucosidase. It undergoes estrification to obtain suitable monomers for emulsion polymerization itaconates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Itaconic anhydride is used in immobilization of β-glucosidase. It undergoes estrification to obtain suitable monomers for emulsion polymerization itaconates.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text