Thermo Scientific Chemicals

1-Bromo-4-phenylbutane, 98%, Thermo Scientific Chemicals

Catalog number: B25237.06
5 g, Each
Thermo Scientific Chemicals

1-Bromo-4-phenylbutane, 98%, Thermo Scientific Chemicals

Catalog number: B25237.06
5 g, Each
Quantity
Catalog number: B25237.06
also known as B25237-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13633-25-5
IUPAC Name
(4-bromobutyl)benzene
Molecular Formula
C10H13Br
InChI Key
XPBQQAHIVODAIC-UHFFFAOYSA-N
SMILES
BrCCCCC1=CC=CC=C1
Refractive Index
1.5390-1.5430 @ 20?C
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%

Description

1-Bromo-4-phenylbutane is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-phenylbutane is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text