Thermo Scientific Chemicals

4-Bromomethyl-3-nitrobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: B25255.06
5 g, Each
Thermo Scientific Chemicals

4-Bromomethyl-3-nitrobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: B25255.06
5 g, Each
Quantity
Catalog number: B25255.06
also known as B25255-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
55715-03-2
IUPAC Name
4-(bromomethyl)-3-nitrobenzoate
Molecular Formula
C8H5BrNO4
InChI Key
QMAHVAFURJBOFV-UHFFFAOYSA-M
SMILES
[O-]C(=O)C1=CC=C(CBr)C(=C1)[N+]([O-])=O
Melting Point (clear melt)
125-137°C
Appearance (Color)
White to cream or pale yellow to pale brown
Form
Crystalline powder or powder
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (HPLC)
≥96.0%

Description

4-Bromomethyl-3-nitrobenzoic acid is used as a reactant in the synthesis of 4-bromomethyl-3-nitrobenzoic acid succinimide ester, 4-((2-(hydroxymethyl)phenyl amino)methyl)-3-nitrobenzoic acid, 4-(2-hydroxyethylmercaptylmethyl)-3-nitrobenzoic acid. It is also used as a thiol photo-deprotection reagent, a starting material in the synthesis of 2H-indazole based library using parallel solution-phase methods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromomethyl-3-nitrobenzoic acid is used as a reactant in the synthesis of 4-bromomethyl-3-nitrobenzoic acid succinimide ester, 4-((2-(hydroxymethyl)phenyl amino)methyl)-3-nitrobenzoic acid, 4-(2-hydroxyethylmercaptylmethyl)-3-nitrobenzoic acid. It is also used as a thiol photo-deprotection reagent, a starting material in the synthesis of 2H-indazole based library using parallel solution-phase methods.

Solubility
Soluble in DMF and dichloromethane. Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text