Methyl 2-oxocyclopentanecarboxylate, 97%, Thermo Scientific Chemicals
Methyl 2-oxocyclopentanecarboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 2-oxocyclopentanecarboxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number B25256.22
also known as B25256-22
Price (USD)/ Each
227.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
227.00
Add to cart
Methyl 2-oxocyclopentanecarboxylate, 97%, Thermo Scientific Chemicals
Catalog numberB25256.22
Price (USD)/ Each
227.00
-
Add to cart
Chemical Identifiers
CAS10472-24-9
IUPAC Namemethyl 2-oxocyclopentane-1-carboxylate
Molecular FormulaC7H10O3
InChI KeyPZBBESSUKAHBHD-UHFFFAOYNA-N
SMILESCOC(=O)C1CCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale pink
Assay (Silylated GC)≥96.0%
FormLiquid
Refractive Index1.4540-1.4590 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only