Thermo Scientific Chemicals

Alginic acid sodium salt, low viscosity, Thermo Scientific Chemicals

Catalog number: B25266.36
500 g, Each
Thermo Scientific Chemicals

Alginic acid sodium salt, low viscosity, Thermo Scientific Chemicals

Catalog number: B25266.36
500 g, Each
Quantity
Catalog number: B25266.36
also known as B25266-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9005-38-3
Molecular Formula
(C6H7O7)A(C6H7O7)BNa
SMILES
[Na+].O[C@H]1[C@@H](O)[C@H](O-*)[C@H](O[C@H]1O-*)C([O-])=O.O[C@H]1[C@@H](O)[C@H](O-*)[C@@H](O[C@H]1O-*)C([O-])=O
Molecular Weight (g/mol)
NaN
Appearance (Color)
White to cream to pale brown or yellowish brown
Loss on Drying
≤13.0%
Total ash
18-27% (dried basis)
pH
5.0-7.5 (1% solution)
Viscosity
30-90 mPas (1% aq. soln.)

Description

A gelling polysaccharide extracted from giant brown seaweed, it is useful in viscosity studiesAlginic acid sodium salt is widely utilized in the food industry as flavorless gum which enhances viscosity and acts as an emulsifier. It finds application as a thickener for reactive dyestuffs which is used in textile screen printing and carpet jet-printing. It is used as indigestion tablets and for the preparation of dental impressions. As a polymer, it combines with chitosan to synthesize a biodegradable porous scaffold which is useful for tissue engineering applications. It acts as a food additive and is used in production of gel-like foods such as baking. It is also used for encapsulating agents of microparticles of beta-galactosidae and in the preparation of alginate hydrogels with tunable mechanical properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text