5-Bromo-2-hydroxypyridine, 97%, Thermo Scientific Chemicals
5-Bromo-2-hydroxypyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromo-2-hydroxypyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B25267.06
also known as B25267-06
Price (USD)/ Each
53.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
53.00
Add to cart
5-Bromo-2-hydroxypyridine, 97%, Thermo Scientific Chemicals
Catalog numberB25267.06
Price (USD)/ Each
53.00
-
Add to cart
Chemical Identifiers
CAS13466-38-1
IUPAC Name5-bromo-1,2-dihydropyridin-2-one
Molecular FormulaC5H4BrNO
InChI KeyNDMZZQRNZFWMEZ-UHFFFAOYSA-N
SMILESBrC1=CNC(=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
FormPowder
Melting Point (clear melt)177.0-186.0?C
Appearance (Color)White to cream to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only