Thermo Scientific Chemicals

3-Nitrophthalonitrile, 99%, Thermo Scientific Chemicals

Catalog number: B25269.06
5 g, Each
Thermo Scientific Chemicals

3-Nitrophthalonitrile, 99%, Thermo Scientific Chemicals

Catalog number: B25269.06
5 g, Each
Quantity
Catalog number: B25269.06
also known as B25269-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51762-67-5
Assay (GC)
≥98.5%
Melting Point (clear melt)
158.0-165.0°C
Form
Powder
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream to pale yellow to black

Description

3-Nitrophthalonitrile is a useful dye for biological research purposes. It is used as an intermediate in the manufacture of pharmaceuticals, chemicals, dyestuff and phthalocyanine blue pigments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitrophthalonitrile is a useful dye for biological research purposes. It is used as an intermediate in the manufacture of pharmaceuticals, chemicals, dyestuff and phthalocyanine blue pigments.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incombatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text