2,4-Dichlorobenzylideneacetone, 97%, Thermo Scientific Chemicals
2,4-Dichlorobenzylideneacetone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-Dichlorobenzylideneacetone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B25270.03
also known as B25270-03
Price (USD)/ Each
23.90
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
23.90
Add to cart
2,4-Dichlorobenzylideneacetone, 97%, Thermo Scientific Chemicals
Catalog numberB25270.03
Price (USD)/ Each
23.90
-
Add to cart
Chemical Identifiers
CAS61888-78-6
IUPAC Name4-(2,4-dichlorophenyl)but-3-en-2-one
Molecular FormulaC10H8Cl2O
InChI KeySAEQHTBHPNKKRX-UHFFFAOYSA-N
SMILESCC(=O)C=CC1=C(Cl)C=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormPowder
Assay (GC)≥96.0%
Melting Point (clear melt)81.5-87.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only