Thermo Scientific Chemicals

1-Tritylimidazole-4-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: B25272.14
25 g, Each
Thermo Scientific Chemicals

1-Tritylimidazole-4-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: B25272.14
25 g, Each
Quantity
Catalog number: B25272.14
also known as B25272-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
33016-47-6
Appearance (Color)
White to pale yellow
Form
Powder
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Free acid (titration)
≤1%

Description

1-Tritylimidazole-4-carboxaldehyde is used in the synthesis of histamine and girolline

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Tritylimidazole-4-carboxaldehyde is used in the synthesis of histamine and girolline

Notes
Air sensitive. Store under inert gas. Incompatible with oxidizing agents, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text