Methyl 2-hydroxy-5-nitrobenzoate, 98%, Thermo Scientific Chemicals
Methyl 2-hydroxy-5-nitrobenzoate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 2-hydroxy-5-nitrobenzoate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number B25278.30
also known as B25278-30
Price (USD)/ Each
614.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
614.00
Add to cart
Methyl 2-hydroxy-5-nitrobenzoate, 98%, Thermo Scientific Chemicals
Catalog numberB25278.30
Price (USD)/ Each
614.00
-
Add to cart
Chemical Identifiers
CAS17302-46-4
IUPAC Name2-(methoxycarbonyl)-4-nitrobenzen-1-olate
Molecular FormulaC8H6NO5
InChI KeyUUBFELFUKFJSRD-UHFFFAOYSA-M
SMILESCOC(=O)C1=CC(=CC=C1[O-])[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to cream to orange
Assay (GC)≥97.5%
Melting Point (clear melt)113.0-119.0?C
FormCrystals or powder or crystalline powder
Methyl 2-hydroxy-5-nitrobenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-hydroxy-5-nitrobenzoate is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. John McGinley; Vickie McKee; Hans Toftlund; John M.D.Walsh. Unusual metal coordination chemistry from an amino-amide derivative of 4-nitrophenol, a surprising ligand. Dalton Trans. 2009, (39), 8406-8412.
  2. David R.Edwards; Alexei A.Neverov; R.Stan Brown. Dissociative solvolytic cleavage of methyl (ortho-carboxymethyl)aryl phosphate diesters mediated by Yb3+ in methanol gives a 1012-fold rate acceleration attributable to leaving group assistance. J. Am. Chem. Soc. 2009, 131,(1), 368-377.