Thermo Scientific Chemicals

2-Amino-3,4-dimethylbenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: B25313.14
25 g, Each
Thermo Scientific Chemicals

2-Amino-3,4-dimethylbenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: B25313.14
25 g, Each
Quantity
Catalog number: B25313.14
also known as B25313-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50419-58-4
Form
Crystals or powder or crystalline powder
Appearance (Color)
Pale brown
Assay (GC)
≥98.0%

Description

2-Amino-3,4-dimethylbenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-3,4-dimethylbenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text